(4,6-Dichloropyrimidin-5-yl)(4-methoxyphenyl)methanone ≥95%

Supplier: Matrix Scientific

072980-250MG 072980-1G
102527-758EA 967.69 USD
102527-758 102527-760
(4,6-Dichloropyrimidin-5-yl)(4-methoxyphenyl)methanone ≥95%
(4,6-Dichloropyrimidin-5-yl)(4-methoxyphenyl)methanone

Hazard: Irritant

Formula: C₁₂H₈Cl₂N₂O₂
MW: 283.11 g/mol
MDL Number: MFCD11109887
CAS Number: 1245646-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR