1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylic acid ethyl ester, Ethyl 1-Boc-4-oxo-3-piperidinecarboxylate, Ethyl 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylate, 1-Boc-3-ethoxycarbonyl-4-piperidone, 1-tert-Butyl 3-ethyl 4-oxopiperidine-1,3-dicarboxylate, 1-Boc-4-oxo-3-piperidinecarboxylic acid ethyl ester, 1-tert-Butoxycarbonyl-3-ethoxycarbonyl-4-piperidone

072978-5G 072978-25G
102527-748EA 218.46 USD
102527-748 102833-578
1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate ≥95%
1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₂₁NO₅
MW: 271.3100 g/mol
Melting Pt: 52…54 °C
MDL Number: MFCD01862189
CAS Number: 98977-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR