(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane ≥95%
Supplier: Matrix Scientific
Synonyms:
(2S,3S)-N-t-Boc-3-amino-1,2-epoxy-4-phenylbutane, (2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, Darunavir Impurity 22
072976-1G
072976-5G
072976-10G
102527-736EA
18.46
USD
102527-736
102527-738
102527-740
(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane ≥95%
(2S,3S)-N-tert-Boc-3-amino-1,2-epoxy-4-phenylbutane
Hazard: Irritant
Formula:
C₁₅H₂₁NO₃ MW: 263.34 g/mol |
MDL Number:
MFCD02258997 CAS Number: 98737-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...