Thiophene-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Thiophenecarboxaldehyde, Thiophene-2-carboxaldehyde

072975-100G 072975-500G
102527-734EA 41.54 USD
102527-734 102663-916
Thiophene-2-carbaldehyde ≥95%
Thiophene-2-carbaldehyde

Hazard: Irritant/Air Sensitive

Formula: C₅H₄OS
MW: 112.15 g/mol
Boiling Pt: 196…198 °C
Density: 1.22
Flash Pt: 78 °C
MDL Number: MFCD00005429
CAS Number: 98-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR