2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-phenyl-5-(trifluoromethyl)pyrazole

072974-1G 072974-5G
102527-728EA 223.08 USD
102527-728 102527-730
2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol ≥95%
2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O
MW: 228.1700 g/mol
MDL Number: MFCD03714773
CAS Number: 96145-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR