5-(Aminomethyl)-2-methylpyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: Grewe diamine, 4-Amino-5-aminomethyl-2-methylpyrimidine

072972-1G 072972-5G
102527-720EA 523.08 USD
102527-720 102527-722
5-(Aminomethyl)-2-methylpyrimidin-4-amine ≥95%
5-(Aminomethyl)-2-methylpyrimidin-4-amine

Hazard: Irritant

Formula: C₆H₁₀N₄
MDL Number: MFCD00078880
CAS Number: 95-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR