Methyl-3-(4-(bromomethyl)phenyl)acrylate ≥98%
Supplier: Matrix Scientific
Synonyms:
Methyl 3-(4-(bromomethyl)phenyl)acrylate, Methyl 4-(bromomethyl)cinnamate
072971-1G
072971-5G
072971-10G
102527-714EA
30.77
USD
102527-714
102527-716
102527-718
Methyl-3-(4-(bromomethyl)phenyl)acrylate ≥98%
Methyl-3-(4-(bromomethyl)phenyl)acrylate
Hazard: Irritant-Harmful/Lachrymator
Formula:
C₁₁H₁₁BrO₂ MW: 255.1100 g/mol Melting Pt: 58…61 °C |
MDL Number:
MFCD02684186 CAS Number: 946-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...