1-Pyridine-4-ylpiperidine-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
072970-1G
072970-5G
102527-710EA
433.85
USD
102527-710
102527-712
1-Pyridine-4-ylpiperidine-4-carboxylic acid ≥95%
1-Pyridine-4-ylpiperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₂ |
CAS Number:
93913-86-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...