Pyrrole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1H-Pyrrole-3-carboxylic acid

072966-1G 072966-5G 072966-10G
102527-690EA 61.54 USD
102527-690 102527-692 102527-694
Pyrrole-3-carboxylic acid ≥95%
Pyrrole-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₅NO₂
MW: 111.10 g/mol
MDL Number: MFCD00800594
CAS Number: 931-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR