1-(2-Amino-4-methoxy-3-methylphenyl)ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 2'-Amino-3'-methyl-4'-methoxyacetophenone

072965-1G 072965-5G 072965-10G
102527-684EA 533.85 USD
102527-684 102527-686 102527-688
1-(2-Amino-4-methoxy-3-methylphenyl)ethanone ≥95%
1-(2-Amino-4-methoxy-3-methylphenyl)ethanone

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MW: 179.2200 g/mol
MDL Number: MFCD11042290
CAS Number: 912347-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR