6-Cyano-1-tetralone ≥95%

Supplier: Matrix Scientific

072964-1G 072964-5G
102527-680EA 353.85 USD
102527-680 102527-682
6-Cyano-1-tetralone ≥95%
6-Cyano-1-tetralone

Hazard: Irritant

Formula: C₁₁H₉NO
MDL Number: MFCD02179289
CAS Number: 90401-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR