5-Chloro-2-hydroxypyrazine ≥95%

Supplier: Matrix Scientific

072962-1G 072962-5G 072962-25G
102527-668EA 153.85 USD
102527-668 102527-670 102551-824
5-Chloro-2-hydroxypyrazine ≥95%
5-Chloro-2-hydroxypyrazine

Hazard: Irritant

Formula: C₄H₃ClN₂O
MW: 130.5300 g/mol
MDL Number: MFCD11109884
CAS Number: 89180-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR