(2,6-Difluoro-4-methoxyphenyl)acetic acid ≥95%

Supplier: Matrix Scientific

072961-1G 072961-5G
102527-662EA 235.38 USD
102527-662 102527-664
(2,6-Difluoro-4-methoxyphenyl)acetic acid ≥95%
(2,6-Difluoro-4-methoxyphenyl)acetic acid

Hazard: Irritant

Formula: (C₆H₂F₂OCH₃)CH₂COOH
MDL Number: MFCD04115927
CAS Number: 886498-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR