2-(3-Amino-5-bromopyridin-2-yl)acetic acid ≥95%

Supplier: Matrix Scientific

072960-250MG 072960-1G
102527-658EA 684.62 USD
102527-658 102527-660
2-(3-Amino-5-bromopyridin-2-yl)acetic acid ≥95%
2-(3-Amino-5-bromopyridin-2-yl)acetic acid

Hazard: Irritant

Formula: C₇H₇BrN₂O₂
CAS Number: 886373-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR