3-Phenylpyridin-2-amine ≥95%

Supplier: Matrix Scientific

072958-250MG 072958-1G
102527-648EA 520.8 USD
102527-648 102527-650
3-Phenylpyridin-2-amine ≥95%
3-Phenylpyridin-2-amine

Hazard: Irritant

Formula: C11H10N2
MW: 170.2100 g/mol
MDL Number: MFCD04114119
CAS Number: 87109-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR