tert-Butyl-3-oxoazepane-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Azepan-3-one, N-BOC protected, N-Boc-3-azacycloheptan-1-one

072957-250MG 072957-1G
102527-644EA 453.77 USD
102527-644 102527-646
tert-Butyl-3-oxoazepane-1-carboxylate ≥95%
tert-Butyl-3-oxoazepane-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MW: 213.2735 g/mol
MDL Number: MFCD08443803
CAS Number: 870842-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR