(R)-3-Amino-1-N-Cbz-pyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

072956-1G 072956-5G 072956-10G
102527-638EA 178.46 USD
102527-638 102527-640 102527-642
(R)-3-Amino-1-N-Cbz-pyrrolidine hydrochloride ≥95%
(R)-3-Amino-1-N-Cbz-pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD09038802
CAS Number: 870621-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR