6-Oxo-1,6-dihydropyridazine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1,6-Dihydro-6-oxopyridazine-4-carboxylic acid

072955-1G 072955-5G
102527-634EA 396.92 USD
102527-634 102527-636
6-Oxo-1,6-dihydropyridazine-4-carboxylic acid ≥95%
6-Oxo-1,6-dihydropyridazine-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₄N₂O₃
MW: 140.0969 g/mol
MDL Number: MFCD08437486
CAS Number: 867130-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR