2-Fluoro-(5-methoxycarbonyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-5-(methoxycarbonyl)phenylboronic acid, (2-Fluoro-5-(methoxycarbonyl)phenyl)boronic acid

072954-1G 072954-5G 072954-10G
102527-626EA 427.69 USD
102527-626 102527-628 102527-630
2-Fluoro-(5-methoxycarbonyl)phenylboronic acid ≥95%
2-Fluoro-(5-methoxycarbonyl)phenylboronic acid

Hazard: Irritant

Formula: C₈H₈BFO₄
MW: 197.9600 g/mol
MDL Number: MFCD06659868
CAS Number: 850568-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR