4-(2-(Piperidin-1-yl)ethoxy)benzoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[2-(1-Piperidyl)ethoxy]benzoic Acid Hydrochloride

072952-1G 072952-5G 072952-10G
102527-614EA 64.62 USD
102527-614 102527-616 102527-618
4-(2-(Piperidin-1-yl)ethoxy)benzoic acid hydrochloride ≥95%
4-(2-(Piperidin-1-yl)ethoxy)benzoic acid hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD06657755
CAS Number: 84449-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR