Methyl-3-ethyl-1H-pyrazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-ethyl-1H-pyrazole-5-carboxylate, 3-Ethyl-5-pyrazolcarboxylic acid methyl ester

072951-250MG 072951-1G 072951-5G
102527-608EA 350.81 USD
102527-608 102527-610 102527-612
Methyl-3-ethyl-1H-pyrazole-5-carboxylate ≥95%
Methyl-3-ethyl-1H-pyrazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MW: 154.1665 g/mol
MDL Number: MFCD08236689
CAS Number: 834869-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR