Ethyl-2-(dimethylaminomethyl)-4-thiazolecarboxylate ≥95%

Supplier: Matrix Scientific

072950-1G 072950-5G
102527-604EA 710.77 USD
102527-604 102527-606
Ethyl-2-(dimethylaminomethyl)-4-thiazolecarboxylate ≥95%
Ethyl-2-(dimethylaminomethyl)-4-thiazolecarboxylate

Hazard: Irritant

Formula: C₉H₁₄N₂O₂S
CAS Number: 82586-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR