3-Amino-N-phenylbenzenesulphonamide ≥95%

Supplier: Matrix Scientific

072949-1G 072949-5G
102527-600EA 355.38 USD
102527-600 102527-602
3-Amino-N-phenylbenzenesulphonamide ≥95%
3-Amino-N-phenylbenzenesulphonamide

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂S
MDL Number: MFCD00035928
CAS Number: 80-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR