Isoquinoline-8-carbaldehyde ≥95%

Supplier: Matrix Scientific

072948-1G 072948-5G
102527-596EA 640 USD
102527-596 102527-598
Isoquinoline-8-carbaldehyde ≥95%
Isoquinoline-8-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₇NO
MW: 157.1687 g/mol
MDL Number: MFCD06227440
CAS Number: 787615-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR