Ethyl-5-bromopicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-bromopicolinate, Ethyl 5-bromopyridine-2-carboxylate

072946-1G 072946-5G
102527-586EA 123.08 USD
102527-586 102527-588
Ethyl-5-bromopicolinate ≥95%
Ethyl 5-bromopicolinate

Hazard: Irritant

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
MDL Number: MFCD00234415
CAS Number: 77199-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR