(S)-2-Methylmorpholine ≥95%

Supplier: Matrix Scientific

072943-250MG 072943-1G
102527-572EA 380 USD
102527-572 102527-574
(S)-2-Methylmorpholine ≥95%
(S)-2-Methylmorpholine

Hazard: Irritant

Formula: C₅H₁₁NO
MW: 101.1469 g/mol
MDL Number: MFCD11506701
CAS Number: 74572-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR