Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

072941-250MG 072941-1G 072941-5G
102527-560EA 355.38 USD
102527-560 102527-562 102527-564
Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-phenyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₁F₃N₂O₂
MW: 284.2339 g/mol
MDL Number: MFCD10568308
CAS Number: 741717-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR