4-Hydroxycinnamic acid ≥95%

Supplier: Matrix Scientific
Warning

Synonyms: p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid

072940-10G 072940-100G
102527-556EA 44.62 USD
102527-556 102527-558
4-Hydroxycinnamic acid ≥95%
4-Hydroxycinnamic acid

Hazard: Irritant

Formula: C₉H₈O₃
MW: 164.16 g/mol
MDL Number: MFCD00004399
CAS Number: 7400-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR