Methyl-2-mercaptobenzo[d]oxazole-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-mercaptobenzo[d]oxazole-6-carboxylate

072939-250MG 072939-1G 072939-5G
102527-550EA 355.38 USD
102527-550 102527-552 102527-554
Methyl-2-mercaptobenzo[d]oxazole-6-carboxylate ≥95%
Methyl-2-mercaptobenzo[d]oxazole-6-carboxylate

Hazard: Irritant

Formula: C₉H₇NO₃S
MW: 209.2218 g/mol
MDL Number: MFCD09261169
CAS Number: 72752-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR