Ethyl-2-(4-(trifluoromethyl)phenyl)acetate ≥95%

Supplier: Matrix Scientific

072938-1G 072938-5G 072938-25G
102527-544EA 36.92 USD
102527-544 102527-546 102833-576
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate ≥95%
Ethyl-2-(4-(trifluoromethyl)phenyl)acetate

Hazard: Irritant

Formula: C₁₁H₁₁F₃O₂
MDL Number: MFCD07779351
CAS Number: 721-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR