2,7-Dihydroxyquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: 2,7-Quinolinediol

072934-1G 072934-5G
102527-522EA 18.46 USD
102527-522 102527-524
2,7-Dihydroxyquinoline ≥95%
2,7-Dihydroxyquinoline

Hazard: Irritant

Formula: C₉H₇NO₂
MW: 161.157 g/mol
Melting Pt: 187-192°C
MDL Number: MFCD07644575
CAS Number: 70500-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR