Methyl-3-cyano-4-fluorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-cyano-4-fluorobenzoate

072926-1G 072926-5G
102527-480EA 387.69 USD
102527-480 102527-482
Methyl-3-cyano-4-fluorobenzoate ≥95%
Methyl-3-cyano-4-fluorobenzoate

Hazard: Irritant

Formula: C₉H₆FNO₂
MW: 179.1500 g/mol
MDL Number: MFCD06203915
CAS Number: 676602-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR