5-(Hydroxymethyl)-2-furaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxymethyl-2-furancarbaldehyde, 5-(hydroxymethyl)furan-2-carbaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethylfurfural

072925-5G 072925-25G
102527-476EA 60 USD
102527-476 102833-572
5-(Hydroxymethyl)-2-furaldehyde ≥95%
5-(Hydroxymethyl)-2-furaldehyde

Hazard: Irritant/Refrigerate/Stored under Argon

Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 114-116°C/1mm
Melting Pt: 32-35°C
Density: 1.206
Flash Pt: 79°C
MDL Number: MFCD00003234
CAS Number: 67-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR