O-(Tetrahydropyran-2-yl)-hydroxylamine ≥95%

Supplier: Matrix Scientific

072923-5G 072923-25G
102527-464EA 53.85 USD
102527-464 102551-822
O-(Tetrahydropyran-2-yl)-hydroxylamine ≥95%
O-(Tetrahydropyran-2-yl)-hydroxylamine

Hazard: Irritant

Formula: C₅H₁₁NO₂
Boiling Pt: 81 °C (20 mmHg)
Melting Pt: 34…37 °C
Flash Pt: 82 °C
MDL Number: MFCD01321374
CAS Number: 6723-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR