(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone ≥95%

Supplier: Matrix Scientific

072921-1G 072921-5G 072921-25G
102527-454EA 23.08 USD
102527-454 102527-456 102663-914
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone ≥95%
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone

Hazard: Irritant

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 83…85 °C
MDL Number: MFCD00077791
CAS Number: 66673-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR