3-Fluoro-4-iodoaniline ≥95%

Supplier: Matrix Scientific

072920-1G 072920-5G
102527-448EA 15.38 USD
102527-448 102527-450
3-Fluoro-4-iodoaniline ≥95%
3-Fluoro-4-iodoaniline

Hazard: Irritant

Formula: C₆H₅FIN
MW: 237.01 g/mol
MDL Number: MFCD07774187
CAS Number: 656-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR