3-Quinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

072916-5G 072916-10G
102527-426EA 53.85 USD
102527-426 102527-428
3-Quinolinecarboxylic acid ≥95%
3-Quinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
MDL Number: MFCD00006770
CAS Number: 6480-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR