tert-Butyl (4-(aminomethyl)pyridin-2-yl)carbamate ≥95%

Supplier: Matrix Scientific

072915-250MG 072915-1G 072915-5G
102527-420EA 427.69 USD
102527-420 102527-422 102527-424
tert-Butyl (4-(aminomethyl)pyridin-2-yl)carbamate ≥95%
tert-Butyl (4-(aminomethyl)pyridin-2-yl)carbamate

Hazard: Irritant

Formula: C₁₁H₁₇N₃O₂
CAS Number: 639091-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR