4,6-Dichloro-2-(methylthio)pyrimidine ≥95%

Supplier: Matrix Scientific

072913-100G 072913-25G
102527-412EA 227.69 USD
102527-412 102551-818
4,6-Dichloro-2-(methylthio)pyrimidine ≥95%
4,6-Dichloro-2-(methylthio)pyrimidine

Hazard: Corrosive

Formula: C₅H₄Cl₂N₂S
MW: 195.07 g/mol
MDL Number: MFCD00006086
CAS Number: 6299-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR