3-Amino-4-fluorophenol ≥95%
Supplier: Matrix Scientific
072911-1G
072911-5G
072911-10G
102527-396EA
33.85
USD
102527-396
102527-398
102527-400
3-Amino-4-fluorophenol ≥95%
3-Amino-4-fluorophenol
Hazard: Irritant
Formula:
C₆H₆FNO MW: 127.1200 g/mol |
MDL Number:
MFCD09033850 CAS Number: 62257-16-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...