2-Chloro-5-nitrophenol ≥95%

Supplier: Matrix Scientific

072910-1G 072910-5G 072910-10G
102527-390EA 72.31 USD
102527-390 102527-392 102527-394
2-Chloro-5-nitrophenol ≥95%
2-Chloro-5-nitrophenol

Hazard: Irritant

Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
MDL Number: MFCD01571825
CAS Number: 619-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR