1,1'-Thiocarbonyldiimidazole ≥95%

Supplier: Matrix Scientific

072909-10G 072909-100G
102527-386EA 143.08 USD
102527-386 102527-388
1,1'-Thiocarbonyldiimidazole ≥95%
1,1'-Thiocarbonyldiimidazole

Hazard: Irritant/Stored under Argon

Formula: C₇H₆N₄S
MDL Number: MFCD00005289
CAS Number: 6160-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR