7-Hydroxy-1,2,3,4-tetrahydroquinoline ≥95%

Supplier: Matrix Scientific

072901-1G 072901-5G
102527-348EA 90.77 USD
102527-348 102527-350
7-Hydroxy-1,2,3,4-tetrahydroquinoline ≥95%
7-Hydroxy-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₉H₁₁NO
MW: 149.1897 g/mol
MDL Number: MFCD08063887
CAS Number: 58196-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR