3-Quinolinol ≥95%

Supplier: Matrix Scientific

072900-1G 072900-5G
102527-344EA 21.54 USD
102527-344 102527-346
3-Quinolinol ≥95%
3-Quinolinol

Hazard: Irritant

Formula: C₉H₇NO
MDL Number: MFCD00169018
CAS Number: 580-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR