6-Quinolinol ≥95%

Supplier: Matrix Scientific

Synonyms: Quinolin-6-ol, 6-Hydroxyquinoline

072899-5G 072899-10G
102527-338EA 107.69 USD
102527-338 102527-340
6-Quinolinol ≥95%
6-Quinolinol

Hazard: Irritant

Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00047611
CAS Number: 580-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR