7-MAC ≥95%

Supplier: Matrix Scientific

Synonyms: Diphenylmethyl (6R,7S)-7-Amino-7-methoxy-3-[(1-methyl-1H-tetrazol-5-yl)thiomethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Diphenylmethyl 7beta-Amino-7alpha-methoxy-3-[(1-methyl-1H-tetrazol-5-yl)thiomethyl]-3-cephem-4-carboxylate, Benzhydryl (6R,7S)-7-amino-7-methoxy-3-(((1-methyl-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

072894-1G 072894-5G 072894-10G
102527-310EA 213.85 USD
102527-310 102527-312 102527-314
7-MAC ≥95%
7-MAC

Hazard: Irritant

Formula: C₂₄H₂₄N₆O₄S₂
MW: 524.6152 g/mol
MDL Number: MFCD08704705
CAS Number: 56610-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR