2,6-Dibromo-3-nitropyridine ≥95%

Supplier: Matrix Scientific

072890-1G 072890-5G
102527-290EA 15.38 USD
102527-290 102527-292
2,6-Dibromo-3-nitropyridine ≥95%
2,6-Dibromo-3-nitropyridine

Hazard: Irritant

Formula: C₅H₂Br₂N₂O₂
MW: 281.8900 g/mol
MDL Number: MFCD00233995
CAS Number: 55304-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR