6-Methylpyrazine-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
072889-1G
072889-5G
102527-286EA
113.85
USD
102527-286
102527-288
6-Methylpyrazine-2-carboxylic acid ≥95%
6-Methylpyrazine-2-carboxylic acid
Hazard: Irritant
Formula:
C₆H₆N₂O₂ MW: 138.1200 g/mol |
MDL Number:
MFCD08705756 CAS Number: 5521-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...