4-Aminopiperidin-2-one hydrochloride ≥95%

Supplier: Matrix Scientific

072888-250MG 072888-1G
102527-282EA 967.69 USD
102527-282 102527-284
4-Aminopiperidin-2-one hydrochloride ≥95%
4-Aminopiperidin-2-one hydrochloride

Hazard: Irritant

Formula: C₅H₁₁ClN₂O
CAS Number: 1260883-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR