3-Ethoxy-2,2-dimethyl-3-oxopropanoic acid ≥95%

Supplier: Matrix Scientific

072886-250MG 072886-1G 072886-5G
102527-270EA 250.11 USD
102527-270 102527-272 102527-274
3-Ethoxy-2,2-dimethyl-3-oxopropanoic acid ≥95%
3-Ethoxy-2,2-dimethyl-3-oxopropanoic acid

Hazard: Irritant

Formula: C7H12O4
MW: 160.1700 g/mol
MDL Number: MFCD04003823
CAS Number: 5471-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR